Product Details

SNP ID
rs183428005
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141100737 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGGGTCTCTGGCTGGGTCCGAG[C/T]CAAGACGCTATTCTGTGGCTGAGGA
Phenotype
MIM: 606328 MIM: 606329
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PCDHB2 PubMed Links

Gene Details

Gene
PCDHB2
Gene Name
protocadherin beta 2
There are no transcripts associated with this gene.

Gene
PCDHB3
Gene Name
protocadherin beta 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018937.4 496 Missense Mutation CCA,TCA P30S NP_061760.1

View Full Product Details