Product Details

SNP ID
rs183380928
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:97191138 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACTGATGAGAAAGATTCTGTTTC[A/G]AACATAGCCACAGAAATAAAGGATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD36 PubMed Links

Gene Details

Gene
ANKRD36
Gene Name
ankyrin repeat domain 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164315.1 2548 Silent Mutation NP_001157787.1
XM_006712514.3 2548 Intron XP_006712577.1
XM_011511130.1 2548 Intron XP_011509432.1
XM_011511131.1 2548 Intron XP_011509433.1
XM_017004009.1 2548 Intron XP_016859498.1
XM_017004010.1 2548 Intron XP_016859499.1
XM_017004011.1 2548 Intron XP_016859500.1
XM_017004012.1 2548 Intron XP_016859501.1
XM_017004013.1 2548 Intron XP_016859502.1
XM_017004014.1 2548 Intron XP_016859503.1
XM_017004015.1 2548 Intron XP_016859504.1
XM_017004016.1 2548 Intron XP_016859505.1
XM_017004017.1 2548 Intron XP_016859506.1
XM_017004018.1 2548 Intron XP_016859507.1
XM_017004019.1 2548 Intron XP_016859508.1
XM_017004020.1 2548 Intron XP_016859509.1
XM_017004021.1 2548 Intron XP_016859510.1
XM_017004022.1 2548 Intron XP_016859511.1
XM_017004023.1 2548 Intron XP_016859512.1
XM_017004024.1 2548 Intron XP_016859513.1
XM_017004025.1 2548 Intron XP_016859514.1
XM_017004026.1 2548 Intron XP_016859515.1
XM_017004027.1 2548 Intron XP_016859516.1
XM_017004028.1 2548 Intron XP_016859517.1
XM_017004029.1 2548 Intron XP_016859518.1
XM_017004030.1 2548 Intron XP_016859519.1
XM_017004031.1 2548 Intron XP_016859520.1
XM_017004032.1 2548 Intron XP_016859521.1
XM_017004033.1 2548 Intron XP_016859522.1
XM_017004034.1 2548 Intron XP_016859523.1
XM_017004035.1 2548 Intron XP_016859524.1
XM_017004036.1 2548 Intron XP_016859525.1
XM_017004037.1 2548 Intron XP_016859526.1
XM_017004038.1 2548 Intron XP_016859527.1
XM_017004039.1 2548 Intron XP_016859528.1
XM_017004040.1 2548 Intron XP_016859529.1
XM_017004041.1 2548 Intron XP_016859530.1
XM_017004042.1 2548 Intron XP_016859531.1
XM_017004043.1 2548 Intron XP_016859532.1
XM_017004044.1 2548 Intron XP_016859533.1
XM_017004045.1 2548 Intron XP_016859534.1
XM_017004046.1 2548 Intron XP_016859535.1
XM_017004047.1 2548 Intron XP_016859536.1
XM_017004048.1 2548 Intron XP_016859537.1
XM_017004049.1 2548 Intron XP_016859538.1
XM_017004050.1 2548 Intron XP_016859539.1
XM_017004051.1 2548 Intron XP_016859540.1

View Full Product Details