Product Details

SNP ID
rs182940784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55014803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCAGGAGGAGGCAGCATGGCCT[C/T]GTTTCCACAGCTGTAGCCTCTGCCC
Phenotype
MIM: 605546
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GP6 PubMed Links

Gene Details

Gene
GP6
Gene Name
glycoprotein VI platelet
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083899.2 1166 Missense Mutation NP_001077368.2
NM_001256017.2 1166 UTR 3 NP_001242946.2
NM_016363.5 1166 UTR 3 NP_057447.5

View Full Product Details