Product Details

SNP ID
rs183975963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:21021485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTACAGGTATTCTAAAGAAACCAA[C/T]ATTGATCCATCAGAAAATTCAACAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLCO1B7 PubMed Links

Gene Details

Gene
SLCO1B7
Gene Name
solute carrier organic anion transporter family member 1B7 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009562.4 243 Silent Mutation AAC,AAT N81N NP_001009562.3

View Full Product Details