Product Details

SNP ID
rs183621422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100401928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGAATTCCCTTGGCCTTCTTTCC[C/T]TCCCATTCTGGGTGCAGGAGGAGCT
Phenotype
MIM: 605341
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PILRA PubMed Links

Gene Details

Gene
PILRA
Gene Name
paired immunoglobin like type 2 receptor alpha
There are no transcripts associated with this gene.

Gene
ZCWPW1
Gene Name
zinc finger CW-type and PWWP domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258008.1 2399 Intron NP_001244937.1
NM_017984.4 2399 Missense Mutation AGG,GGG R529G NP_060454.3
XM_005250479.2 2399 Intron XP_005250536.1
XM_005250480.2 2399 Intron XP_005250537.1
XM_006716035.3 2399 Missense Mutation AGG,GGG R531G XP_006716098.1
XM_006716036.2 2399 Missense Mutation AGG,GGG R531G XP_006716099.1
XM_006716037.3 2399 Missense Mutation AGG,GGG R530G XP_006716100.1
XM_006716038.3 2399 Missense Mutation AGG,GGG R530G XP_006716101.1
XM_006716040.3 2399 Missense Mutation AGG,GGG R504G XP_006716103.1
XM_017012379.1 2399 Intron XP_016867868.1
XM_017012380.1 2399 Intron XP_016867869.1

View Full Product Details