Product Details
- SNP ID
-
rs183729618
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:43361192 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATGACACCCATAGGGTTTCTCTC[C/T]TGTGTGGGTTCTATGATGTGCACGT
- Phenotype
-
MIM: 609610
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADAL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs35278805] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADAL
- Gene Name
- adenosine deaminase like
There are no transcripts associated with this gene.
- Gene
- TUBGCP4
- Gene Name
- tubulin gamma complex associated protein 4
There are no transcripts associated with this gene.
- Gene
- ZSCAN29
- Gene Name
- zinc finger and SCAN domain containing 29
View Full Product Details