Product Details

SNP ID
rs184147813
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74362108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTCAGTGGGTACTGTGGCTCCAG[C/G]GCGCTGAGATACTGTCTCCTTGAGG
Phenotype
MIM: 601143
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DCTN1 PubMed Links

Gene Details

Gene
DCTN1
Gene Name
dynactin subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135040.2 3609 Missense Mutation CCT,GCT P1190A NP_001128512.1
NM_001135041.2 3609 Missense Mutation CCT,GCT P1076A NP_001128513.1
NM_001190836.1 3609 Missense Mutation CCT,GCT P1173A NP_001177765.1
NM_001190837.1 3609 Missense Mutation CCT,GCT P1208A NP_001177766.1
NM_004082.4 3609 Missense Mutation CCT,GCT P1215A NP_004073.2
NM_023019.3 3609 Missense Mutation CCT,GCT P1081A NP_075408.1

View Full Product Details