Product Details

SNP ID
rs184037690
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64193140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTGGGTAACATCGATGATGCCTT[A/G]CAGTGCTACTCCGAAGCTATTAAGC
Phenotype
MIM: 605063
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STIP1 PubMed Links

Gene Details

Gene
STIP1
Gene Name
stress induced phosphoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282652.1 766 Silent Mutation TTA,TTG L71L NP_001269581.1
NM_001282653.1 766 Intron NP_001269582.1
NM_006819.2 766 Intron NP_006810.1

View Full Product Details