Product Details

SNP ID
rs183989443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:131818088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCCTGCTTAGAAAGCTTTTTTT[G/T]ACGTTAGCAAGCATCTGGGGGATCT
Phenotype
MIM: 173335
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ENPP1 PubMed Links

Gene Details

Gene
ENPP1
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006208.2 Intron NP_006199.2
XM_011535896.1 Intron XP_011534198.1

View Full Product Details