Product Details

SNP ID
rs185495405
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:81987657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATTCACAACAGATAGGTGATAG[C/T]TGGGAATGGAGACCATCAAAGGTAA
Phenotype
MIM: 610800
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATG10 PubMed Links

Gene Details

Gene
ATG10
Gene Name
autophagy related 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001131028.1 385 Silent Mutation AGC,AGT S29S NP_001124500.1
NM_031482.4 385 Silent Mutation AGC,AGT S29S NP_113670.1
XM_005248610.4 385 Silent Mutation AGC,AGT S29S XP_005248667.1
XM_005248611.4 385 Intron XP_005248668.1
XM_005248612.3 385 Silent Mutation AGC,AGT S29S XP_005248669.1
XM_011543660.2 385 Intron XP_011541962.1
XM_011543661.2 385 Intron XP_011541963.1
XM_017009944.1 385 Silent Mutation AGC,AGT S29S XP_016865433.1

View Full Product Details