Product Details
- SNP ID
-
rs185305653
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:28326427 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACACACTTTGCATTCATATGGTTT[C/T]TCCCCTGTGTGGATTCTTCTGTGTC
- Phenotype
-
MIM: 610794
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZSCAN31
PubMed Links
Gene Details
- Gene
- ZSCAN31
- Gene Name
- zinc finger and SCAN domain containing 31
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001135215.1 |
1196 |
Silent Mutation |
GAA,GAG |
E320E |
NP_001128687.1 |
NM_001135216.1 |
1196 |
Silent Mutation |
GAA,GAG |
E320E |
NP_001128688.1 |
NM_001243241.1 |
1196 |
Silent Mutation |
GAA,GAG |
E320E |
NP_001230170.1 |
NM_001243242.1 |
1196 |
Silent Mutation |
GAA,GAG |
E161E |
NP_001230171.1 |
NM_001243243.1 |
1196 |
Silent Mutation |
GAA,GAG |
E161E |
NP_001230172.1 |
NM_001243244.1 |
1196 |
Silent Mutation |
GAA,GAG |
E161E |
NP_001230173.1 |
NM_030899.4 |
1196 |
Silent Mutation |
GAA,GAG |
E320E |
NP_112161.3 |
NM_145909.2 |
1196 |
Silent Mutation |
GAA,GAG |
E320E |
NP_665916.1 |
XM_005249295.1 |
1196 |
Silent Mutation |
GAA,GAG |
E320E |
XP_005249352.1 |
XM_005249296.4 |
1196 |
Silent Mutation |
GAA,GAG |
E320E |
XP_005249353.1 |
XM_011514807.2 |
1196 |
Silent Mutation |
GAA,GAG |
E320E |
XP_011513109.1 |
XM_011514808.2 |
1196 |
Silent Mutation |
GAA,GAG |
E320E |
XP_011513110.1 |
XM_011514809.1 |
1196 |
Silent Mutation |
GAA,GAG |
E320E |
XP_011513111.1 |
XM_011514811.2 |
1196 |
Silent Mutation |
GAA,GAG |
E320E |
XP_011513113.1 |
XM_011514812.2 |
1196 |
Silent Mutation |
GAA,GAG |
E320E |
XP_011513114.1 |
XM_011514813.2 |
1196 |
Silent Mutation |
GAA,GAG |
E320E |
XP_011513115.1 |
XM_017011196.1 |
1196 |
Silent Mutation |
GAA,GAG |
E320E |
XP_016866685.1 |
View Full Product Details