Product Details

SNP ID
hCV181730568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:77039519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCTGCCGGTGTCAAAGACGACT[A/T]CCGGCTCACCTCTGACTCGGTTTCC
Phenotype
MIM: 615315
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
B3GNT6 PubMed Links

Gene Details

Gene
B3GNT6
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6
There are no transcripts associated with this gene.

View Full Product Details