Product Details

SNP ID
rs186499615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85609567 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCGGACTCTTCATAGTCTGGGTT[A/G]CGTGGGTGGGCAGAGACATCTGGAA
Phenotype
MIM: 616757 MIM: 611594
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2orf68 PubMed Links

Gene Details

Gene
C2orf68
Gene Name
chromosome 2 open reading frame 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013649.3 591 Silent Mutation CGC,CGT R82R NP_001013671.2
XM_005264305.3 591 UTR 3 XP_005264362.1
Gene
TMEM150A
Gene Name
transmembrane protein 150A
There are no transcripts associated with this gene.

Gene
USP39
Gene Name
ubiquitin specific peptidase 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256725.1 591 Intron NP_001243654.1
NM_001256726.1 591 Intron NP_001243655.1
NM_001256727.1 591 Intron NP_001243656.1
NM_001256728.1 591 Intron NP_001243657.1
NM_006590.3 591 Intron NP_006581.2
XM_006711922.1 591 Intron XP_006711985.1
XM_006711923.2 591 Intron XP_006711986.1
XM_011532487.1 591 Intron XP_011530789.1
XM_011532488.1 591 Intron XP_011530790.1
XM_017003182.1 591 Intron XP_016858671.1

View Full Product Details