Product Details

SNP ID
rs186215242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:34630322 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAGACTATTGGTTCTAGTAGTA[A/G]TCTTCATAGTTCTTAGGGTTCAGGC
Phenotype
MIM: 300698
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM47 PubMed Links

Gene Details

Gene
TMEM47
Gene Name
transmembrane protein 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031442.3 796 Silent Mutation GAC,GAT D179D NP_113630.1

View Full Product Details