Product Details

SNP ID
rs186183930
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:163609975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTACTACGATTTGGTCCTCAGC[C/T]GAAGTTAGTACAGTGTCCAGTTGCA
Phenotype
MIM: 605295
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FIGN PubMed Links

Gene Details

Gene
FIGN
Gene Name
fidgetin, microtubule severing factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321825.1 2172 Silent Mutation NP_001308754.1
NM_018086.3 2172 Silent Mutation NP_060556.2
XM_011511389.2 2172 Silent Mutation XP_011509691.1
XM_017004419.1 2172 Silent Mutation XP_016859908.1

View Full Product Details