Product Details

SNP ID
rs187115849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:4860009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTTTGGTGATGGAGAAATGGCA[A/G]GCTGAGAGACAGCGTGAGGAATACT
Phenotype
MIM: 607436 MIM: 611491
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAPOLB PubMed Links

Gene Details

Gene
PAPOLB
Gene Name
poly(A) polymerase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020144.4 1986 Missense Mutation CCT,CTT P601L NP_064529.4
Gene
RADIL
Gene Name
Rap associating with DIL domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018059.4 1986 Intron NP_060529.4

View Full Product Details