Product Details
- SNP ID
-
rs187394342
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:4968808 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTTCCAGCTCCTGGTTCTGCTTC[A/C]GTTCCCTCATCCTGCAGAGAGAAAG
- Phenotype
-
MIM: 611508
MIM: 610056
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CAMTA2
PubMed Links
Gene Details
- Gene
- CAMTA2
- Gene Name
- calmodulin binding transcription activator 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001171166.1 |
4341 |
Missense Mutation |
CGG,CTG |
R1181L |
NP_001164637.1 |
NM_001171167.1 |
4341 |
Silent Mutation |
ACG,ACT |
T1208T |
NP_001164638.1 |
NM_001171168.1 |
4341 |
Missense Mutation |
CGG,CTG |
R1185L |
NP_001164639.1 |
NM_015099.3 |
4341 |
Missense Mutation |
CGG,CTG |
R1186L |
NP_055914.2 |
XM_006721478.3 |
4341 |
Missense Mutation |
CGG,CTG |
R1191L |
XP_006721541.1 |
XM_006721481.3 |
4341 |
Missense Mutation |
CGG,CTG |
R1162L |
XP_006721544.1 |
XM_006721482.3 |
4341 |
Missense Mutation |
CGG,CTG |
R999L |
XP_006721545.1 |
XM_011523746.2 |
4341 |
Missense Mutation |
CGG,CTG |
R1214L |
XP_011522048.1 |
XM_011523747.2 |
4341 |
Missense Mutation |
CGG,CTG |
R1209L |
XP_011522049.1 |
XM_011523748.2 |
4341 |
Missense Mutation |
CGG,CTG |
R1207L |
XP_011522050.1 |
XM_011523749.2 |
4341 |
Missense Mutation |
CGG,CTG |
R1202L |
XP_011522051.1 |
- Gene
- MIR6864
- Gene Name
- microRNA 6864
There are no transcripts associated with this gene.
- Gene
- MIR6865
- Gene Name
- microRNA 6865
There are no transcripts associated with this gene.
- Gene
- SPAG7
- Gene Name
- sperm associated antigen 7
View Full Product Details