Product Details

SNP ID
rs188726674
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:222749538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCCAAATATCAGTATGTGTTAAA[C/T]GAGTTCTATAGGATACAAAACAAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM177B PubMed Links

Gene Details

Gene
FAM177B
Gene Name
family with sequence similarity 177 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324080.1 1106 Silent Mutation AAC,AAT N105N NP_001311009.1
NM_207468.2 1106 Silent Mutation AAC,AAT N105N NP_997351.2
XM_017001279.1 1106 Silent Mutation AAC,AAT N105N XP_016856768.1
XM_017001280.1 1106 Silent Mutation AAC,AAT N105N XP_016856769.1
XM_017001281.1 1106 Silent Mutation AAC,AAT N105N XP_016856770.1
XM_017001282.1 1106 Silent Mutation AAC,AAT N105N XP_016856771.1
XM_017001283.1 1106 UTR 3 XP_016856772.1
XM_017001284.1 1106 UTR 3 XP_016856773.1
XM_017001285.1 1106 Intron XP_016856774.1

View Full Product Details