Product Details

SNP ID
rs192033192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:98199448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACACAGCTCTGCGCCGGGATGCC[C/T]GCCTCTCCAGGTACTCTGCCTTAAG
Phenotype
MIM: 602159 MIM: 609871
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CORO2A PubMed Links

Gene Details

Gene
CORO2A
Gene Name
coronin 2A
There are no transcripts associated with this gene.

Gene
TBC1D2
Gene Name
TBC1 domain family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267571.1 2900 Missense Mutation CAG,CGG Q907R NP_001254500.1
NM_001267572.1 2900 Missense Mutation CAG,CGG Q447R NP_001254501.1
NM_018421.3 2900 Missense Mutation CAG,CGG Q896R NP_060891.3
XM_011518843.1 2900 Missense Mutation CAG,CGG Q447R XP_011517145.1
XM_011518844.2 2900 Intron XP_011517146.1
XM_017014899.1 2900 Missense Mutation CAG,CGG Q462R XP_016870388.1

View Full Product Details