Product Details

SNP ID
rs192670888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44327499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAGGGTAGTCCTTACCATAGTC[C/T]TCGCTTTTATAGAATTTATCACTAC
Phenotype
MIM: 603994
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF112 PubMed Links

Gene Details

Gene
ZNF112
Gene Name
zinc finger protein 112
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083335.1 2789 Silent Mutation GAA,GAG E892E NP_001076804.1
NM_013380.3 2789 Silent Mutation GAA,GAG E886E NP_037512.3

View Full Product Details