Product Details
- SNP ID
-
rs192685142
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52790127 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTGCTGGCCGAGCTGAAGCCCCCG[C/T]CACTGACTCCATAGCGGGCGCCAGA
- Phenotype
-
MIM: 148043
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KRT3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs570613061] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KRT3
- Gene Name
- keratin 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_057088.2 |
1868 |
Missense Mutation |
GAC,GGC |
D601G |
NP_476429.2 |
View Full Product Details