Product Details

SNP ID
rs192607423
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56988949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAACAGCACTGAAGTCACTGAATT[C/T]TATCTTCTGGGATTTGGTGCCCAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5AK2 PubMed Links

Gene Details

Gene
OR5AK2
Gene Name
olfactory receptor family 5 subfamily AK member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005323.1 36 Silent Mutation TTC,TTT F12F NP_001005323.1

View Full Product Details