Product Details

SNP ID
rs192590104
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:53088059 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTGGATGGTGCAGTGGTTCAGAC[A/T]CGCAGCGGGCAGCTCCTCATCCTGC
Phenotype
MIM: 608661 MIM: 604471
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PODN PubMed Links

Gene Details

Gene
PODN
Gene Name
podocan
There are no transcripts associated with this gene.

Gene
SLC1A7
Gene Name
solute carrier family 1 member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287595.1 1496 Missense Mutation AGT,TGT S604C NP_001274524.1
NM_001287596.1 1496 Intron NP_001274525.1
NM_001287597.1 1496 UTR 3 NP_001274526.1
NM_006671.5 1496 Missense Mutation AGT,TGT S545C NP_006662.3
XM_011542002.2 1496 Missense Mutation AGT,TGT S322C XP_011540304.1

View Full Product Details