Product Details

SNP ID
rs192494277
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1979819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCCTAGGGACGGGCCTCCCTCC[C/G]GGCCTTGGCCCGGGGCCGCCTCCTC
Phenotype
MIM: 600924 MIM: 611256 MIM: 605915
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GFER PubMed Links

Gene Details

Gene
GFER
Gene Name
growth factor, augmenter of liver regeneration
There are no transcripts associated with this gene.

Gene
NOXO1
Gene Name
NADPH oxidase organizer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267721.1 1053 Missense Mutation CCG,CGG P228R NP_001254650.1
NM_144603.3 1053 Missense Mutation CCG,CGG P223R NP_653204.1
NM_172167.2 1053 Missense Mutation CCG,CGG P224R NP_751907.1
NM_172168.2 1053 Missense Mutation CCG,CGG P229R NP_751908.1
XM_005255099.4 1053 Missense Mutation CCG,CGG P229R XP_005255156.1
XM_017022927.1 1053 Missense Mutation CCG,CGG P229R XP_016878416.1
XM_017022928.1 1053 Missense Mutation CCG,CGG P229R XP_016878417.1
Gene
TBL3
Gene Name
transducin beta like 3
There are no transcripts associated with this gene.

View Full Product Details