Product Details

SNP ID
rs5993479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19047320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCCACCCAGCAGCCCCCCAGTG[C/T]GGAGGGACACTGACAACATTCCACT
Phenotype
MIM: 600594
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DGCR11 PubMed Links

Gene Details

Gene
DGCR11
Gene Name
DiGeorge syndrome critical region gene 11 (non-protein coding)
There are no transcripts associated with this gene.

Gene
DGCR2
Gene Name
DiGeorge syndrome critical region gene 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173533.1 Intron NP_001167004.1
NM_001173534.1 Intron NP_001167005.1
NM_001184781.1 Intron NP_001171710.1
NM_005137.2 Intron NP_005128.1

View Full Product Details