Product Details

SNP ID
rs6814166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:106316238 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCCCTCAGCCTGGCCTTTTCTTC[C/T]GCCCTCACAGCCACCGCGACCCAAC
Phenotype
MIM: 603605 MIM: 616899
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AIMP1 PubMed Links

Gene Details

Gene
AIMP1
Gene Name
aminoacyl tRNA synthetase complex interacting multifunctional protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142415.1 29 Intron NP_001135887.1
NM_001142416.1 29 Intron NP_001135888.1
NM_004757.3 29 UTR 5 NP_004748.2
XM_017008835.1 29 Intron XP_016864324.1
XM_017008836.1 29 Intron XP_016864325.1
Gene
TBCK
Gene Name
TBC1 domain containing kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163435.2 29 UTR 5 NP_001156907.1
NM_001163436.2 29 Intron NP_001156908.1
NM_001163437.2 29 UTR 5 NP_001156909.1
NM_001290768.1 29 UTR 5 NP_001277697.1
NM_033115.4 29 UTR 5 NP_149106.2
XM_011532417.2 29 Intron XP_011530719.1
XM_017008846.1 29 UTR 5 XP_016864335.1
XM_017008847.1 29 Intron XP_016864336.1
XM_017008848.1 29 Intron XP_016864337.1
XM_017008849.1 29 Intron XP_016864338.1

View Full Product Details