Product Details

SNP ID
rs7204319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:367551 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGAAGGTGCATGGGCAGCACA[C/T]GAAACCAGGATCCACCCACTGCCCA
Phenotype
MIM: 604853
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL28 PubMed Links

Gene Details

Gene
MRPL28
Gene Name
mitochondrial ribosomal protein L28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006428.4 928 UTR 3 NP_006419.2
XM_005255041.1 928 UTR 3 XP_005255098.1
XM_011522351.2 928 UTR 3 XP_011520653.1
Gene
TMEM8A
Gene Name
transmembrane protein 8A
There are no transcripts associated with this gene.

View Full Product Details