Product Details

SNP ID
rs7355042
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:58576019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGAGAAAGAAAGGAGACCCTGAG[A/G]CCAGGATCTATTAAACCTGGTGTGT
Phenotype
MIM: 137290
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TACSTD2 PubMed Links

Gene Details

Gene
TACSTD2
Gene Name
tumor-associated calcium signal transducer 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002353.2 1476 UTR 3 NP_002344.2

View Full Product Details