Product Details
- SNP ID
-
rs7538082
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
21
- Location
-
Chr.1:18907875 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCATAGTAGTTTAAACACAGTGAG[C/G]CTCTGCCTTCCCCTCACAGTGCCTT
- Phenotype
-
MIM: 606811
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ALDH4A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144039162] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALDH4A1
- Gene Name
- aldehyde dehydrogenase 4 family member A1
There are no transcripts associated with this gene.
- Gene
- IFFO2
- Gene Name
- intermediate filament family orphan 2
View Full Product Details