Product Details

SNP ID
rs7967536
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52013856 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCTCAGCCTGTGGCTCACTCAG[C/G]CTTTGATTCTCCAACCTCAGACTGG
Phenotype
MIM: 612027 MIM: 139139
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GRASP PubMed Links

Gene Details

Gene
GRASP
Gene Name
general receptor for phosphoinositides 1 associated scaffold protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271856.1 Intron NP_001258785.1
NM_181711.3 Intron NP_859062.1
XM_005268691.3 Intron XP_005268748.1
XM_011537996.1 Intron XP_011536298.1
Gene
NR4A1
Gene Name
nuclear receptor subfamily 4 group A member 1
There are no transcripts associated with this gene.

View Full Product Details