Product Details

SNP ID
rs804986
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:120597303 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTGAATTATAGTTTCCTGGTTTC[A/G]TATATAAATGATGTGCATTATGAGG
Phenotype
MIM: 603840
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NDUFB4 PubMed Links

Gene Details

Gene
NDUFB4
Gene Name
NADH:ubiquinone oxidoreductase subunit B4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168331.1 Intron NP_001161803.1
NM_004547.5 Intron NP_004538.2

View Full Product Details