Product Details

SNP ID
rs201255786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:53806747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCAGCCTGTTCCTTAGTGGCTT[C/T]ACCGCTGTATTGTCAGTCCCCACAG
Phenotype
MIM: 605514
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCDH15 PubMed Links

Gene Details

Gene
PCDH15
Gene Name
protocadherin related 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142763.1 5276 Intron NP_001136235.1
NM_001142764.1 5276 Intron NP_001136236.1
NM_001142765.1 5276 Intron NP_001136237.1
NM_001142766.1 5276 Intron NP_001136238.1
NM_001142767.1 5276 Intron NP_001136239.1
NM_001142768.1 5276 Intron NP_001136240.1
NM_001142769.1 5276 Intron NP_001136241.1
NM_001142770.1 5276 Intron NP_001136242.1
NM_001142771.1 5276 Silent Mutation GTA,GTG V1627V NP_001136243.1
NM_001142772.1 5276 Silent Mutation GTA,GTG V1622V NP_001136244.1
NM_001142773.1 5276 Intron NP_001136245.1
NM_033056.3 5276 Intron NP_149045.3
XM_017016571.1 5276 Silent Mutation GTA,GTG V1663V XP_016872060.1
XM_017016572.1 5276 Silent Mutation GTA,GTG V1620V XP_016872061.1
XM_017016573.1 5276 Intron XP_016872062.1

View Full Product Details