Product Details

SNP ID
rs202024115
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5766082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACTCTGATCCTGTCATCCTCTTA[A/T]AGATGTTTTTAATGTCATCACACGT
Phenotype
MIM: 600767
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM208B PubMed Links

Gene Details

Gene
FAM208B
Gene Name
family with sequence similarity 208 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321783.1 1248 Intron NP_001308712.1
NM_001321784.1 1248 Intron NP_001308713.1
NM_001321785.1 1248 Intron NP_001308714.1
NM_017782.4 1248 Intron NP_060252.4
Gene
GDI2
Gene Name
GDP dissociation inhibitor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001115156.1 1248 Missense Mutation TAT,TTT Y376F NP_001108628.1
NM_001494.3 1248 Missense Mutation TAT,TTT Y421F NP_001485.2
XM_017016071.1 1248 Missense Mutation TAT,TTT Y399F XP_016871560.1

View Full Product Details