Product Details

SNP ID
rs202192219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101009329 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCATGGGCTGCACCTTGGACTCA[A/G]TGCCCCCAGAAATGCTGATACCTAG
Phenotype
MIM: 610454 MIM: 612971
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LZTS2 PubMed Links

Gene Details

Gene
LZTS2
Gene Name
leucine zipper tumor suppressor 2
There are no transcripts associated with this gene.

Gene
PDZD7
Gene Name
PDZ domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195263.1 2861 Missense Mutation ACT,ATT T880I NP_001182192.1
NM_024895.4 2861 Intron NP_079171.1
XM_005270165.2 2861 Intron XP_005270222.1
XM_011540177.2 2861 Missense Mutation ACT,ATT T880I XP_011538479.1
XM_011540178.2 2861 Missense Mutation ACT,ATT T879I XP_011538480.1
XM_011540179.2 2861 Intron XP_011538481.1
XM_011540181.2 2861 Intron XP_011538483.1
XM_011540182.2 2861 Intron XP_011538484.1
XM_011540183.2 2861 Intron XP_011538485.1
XM_017016667.1 2861 Intron XP_016872156.1
XM_017016668.1 2861 Intron XP_016872157.1

View Full Product Details