Product Details

SNP ID
rs201443990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73634017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGGCTCGCTCCCATGGGGAAATA[C/T]AGGTCTTGAACACAGTGATATGTTT
Phenotype
MIM: 605603
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYOZ1 PubMed Links

Gene Details

Gene
MYOZ1
Gene Name
myozenin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021245.3 916 Missense Mutation TAT,TGT Y184C NP_067068.1
Gene
USP54
Gene Name
ubiquitin specific peptidase 54
There are no transcripts associated with this gene.

View Full Product Details