Product Details

SNP ID
rs200822371
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:86306346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGGTTTTTATGCTGGGAACAAT[C/T]CCATTTCCTGAGGGAATGGGAGGAT
Phenotype
MIM: 614908
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HIKESHI PubMed Links

Gene Details

Gene
HIKESHI
Gene Name
Hikeshi, heat shock protein nuclear import factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322404.1 147 Silent Mutation ATC,ATT I44I NP_001309333.1
NM_001322407.1 147 Silent Mutation ATC,ATT I5I NP_001309336.1
NM_001322409.1 147 Silent Mutation ATC,ATT I5I NP_001309338.1
NM_016401.3 147 Silent Mutation ATC,ATT I44I NP_057485.2
XM_017017914.1 147 Silent Mutation ATC,ATT I44I XP_016873403.1
XM_017017915.1 147 Silent Mutation ATC,ATT I5I XP_016873404.1

View Full Product Details