Product Details

SNP ID
rs200225575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:30010855 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTTCCATCTCTAGATACTCTGA[C/G]TTGTCCCCCAGGGAAGAAGAAGTAG
Phenotype
MIM: 176266
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KCNA4 PubMed Links

Gene Details

Gene
KCNA4
Gene Name
potassium voltage-gated channel subfamily A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002233.3 3065 Missense Mutation AAC,AAG N608K NP_002224.1

View Full Product Details