Product Details

SNP ID
rs200216663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67433167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCGCAGTGGCCACAACCGGGCTG[C/T]GGACTGGTGGAGCCTGGGGGCCCTG
Phenotype
MIM: 613368 MIM: 609849 MIM: 601577 MIM: 608939
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CARNS1 PubMed Links

Gene Details

Gene
CARNS1
Gene Name
carnosine synthase 1
There are no transcripts associated with this gene.

Gene
CORO1B
Gene Name
coronin 1B
There are no transcripts associated with this gene.

Gene
PTPRCAP
Gene Name
protein tyrosine phosphatase, receptor type C associated protein
There are no transcripts associated with this gene.

Gene
RPS6KB2
Gene Name
ribosomal protein S6 kinase B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003952.2 779 Missense Mutation GCG,GTG A250V NP_003943.2
XM_005274164.1 779 Missense Mutation GCG,GTG A153V XP_005274221.1
XM_005274165.4 779 Missense Mutation GCG,GTG A50V XP_005274222.1
XM_006718655.3 779 Missense Mutation GCG,GTG A50V XP_006718718.1
XM_006718656.3 779 Missense Mutation GCG,GTG A50V XP_006718719.1
XM_006718657.1 779 Missense Mutation GCG,GTG A50V XP_006718720.1
XM_017018108.1 779 Missense Mutation GCG,GTG A50V XP_016873597.1

View Full Product Details