Product Details

SNP ID
rs200669966
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56989434 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCCTTCTGCAAGTCCAATAGCA[C/T]CAATCACTTTTTCTGTGATGTTCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5AK2 PubMed Links

Gene Details

Gene
OR5AK2
Gene Name
olfactory receptor family 5 subfamily AK member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005323.1 521 Missense Mutation ACC,ATC T174I NP_001005323.1

View Full Product Details