Product Details

SNP ID
rs200559511
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:17736130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGCTGGCGGAGCCCGACGCCC[A/C]CAGCCACTTCGACTATGACCCGCGT
Phenotype
MIM: 176258
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KCNC1 PubMed Links

Gene Details

Gene
KCNC1
Gene Name
potassium voltage-gated channel subfamily C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001112741.1 183 Missense Mutation CAC,CCC H43P NP_001106212.1
NM_004976.4 183 Missense Mutation CAC,CCC H43P NP_004967.1
XM_011520080.2 183 Missense Mutation CAC,CCC H43P XP_011518382.1

View Full Product Details