Product Details

SNP ID
rs200696044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58358609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGTATCTCCAATTTGGATAGAA[G/T]CATTCAGAAAACCAATGACATAACA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR5B17 PubMed Links

Gene Details

Gene
OR5B17
Gene Name
olfactory receptor family 5 subfamily B member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005489.1 461 Missense Mutation NP_001005489.1

View Full Product Details