Product Details

SNP ID
rs199919249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63974711 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCCTGACGCCGCTGCTGCTGCGG[A/G]GCTTGACAGGCTCGGCCCGGCGGCT
Phenotype
MIM: 123870
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COX8A PubMed Links

Gene Details

Gene
COX8A
Gene Name
cytochrome c oxidase subunit 8A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004074.2 105 Missense Mutation AGC,GGC S11G NP_004065.1

View Full Product Details