Product Details

SNP ID
rs202177146
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:30011518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGGACAAGCAAAGCAGCGAAC[A/C]ACAAACTCAAAGGAAAACCATACAA
Phenotype
MIM: 176266
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
KCNA4 PubMed Links

Gene Details

Gene
KCNA4
Gene Name
potassium voltage-gated channel subfamily A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002233.3 2402 Silent Mutation GTG,GTT V387V NP_002224.1

View Full Product Details