Product Details

SNP ID
rs201590338
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:9141984 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCTGAAACTTGCCATCCTTGCC[A/G]ATGTTCCGGGGAGTATTGTTGATTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DENND5A PubMed Links

Gene Details

Gene
DENND5A
Gene Name
DENN domain containing 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243254.1 2156 Silent Mutation ATC,ATT I1212I NP_001230183.1
NM_015213.3 2156 Silent Mutation ATC,ATT I1212I NP_056028.2
XM_005252832.1 2156 Silent Mutation ATC,ATT I1212I XP_005252889.1
XM_011519953.2 2156 Silent Mutation ATC,ATT I578I XP_011518255.1
XM_017017419.1 2156 Intron XP_016872908.1

View Full Product Details