Product Details
- SNP ID
-
rs201757557
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:47663074 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGAATCAGGGTGGCCATTGATGG[A/G]TCCAGGGGGGTGTCTTTGTCTCTTC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AGBL2
PubMed Links
Gene Details
- Gene
- AGBL2
- Gene Name
- ATP/GTP binding protein like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024783.3 |
2289 |
Silent Mutation |
GAC,GAT |
D829D |
NP_079059.2 |
XM_005253138.4 |
2289 |
Silent Mutation |
GAC,GAT |
D791D |
XP_005253195.1 |
XM_005253139.4 |
2289 |
Silent Mutation |
GAC,GAT |
D755D |
XP_005253196.1 |
XM_005253140.4 |
2289 |
Silent Mutation |
GAC,GAT |
D713D |
XP_005253197.1 |
XM_005253141.4 |
2289 |
Intron |
|
|
XP_005253198.1 |
XM_006718327.3 |
2289 |
Silent Mutation |
GAC,GAT |
D773D |
XP_006718390.1 |
XM_006718328.3 |
2289 |
Silent Mutation |
GAC,GAT |
D737D |
XP_006718391.1 |
XM_011520372.2 |
2289 |
Silent Mutation |
GAC,GAT |
D791D |
XP_011518674.1 |
XM_011520373.2 |
2289 |
Silent Mutation |
GAC,GAT |
D791D |
XP_011518675.1 |
XM_011520374.2 |
2289 |
Silent Mutation |
GAC,GAT |
D755D |
XP_011518676.1 |
XM_011520375.2 |
2289 |
Silent Mutation |
GAC,GAT |
D746D |
XP_011518677.1 |
XM_011520376.2 |
2289 |
Silent Mutation |
GAC,GAT |
D693D |
XP_011518678.1 |
XM_011520377.2 |
2289 |
Silent Mutation |
GAC,GAT |
D693D |
XP_011518679.1 |
XM_011520378.2 |
2289 |
Silent Mutation |
GAC,GAT |
D693D |
XP_011518680.1 |
XM_011520379.2 |
2289 |
Silent Mutation |
GAC,GAT |
D693D |
XP_011518681.1 |
XM_011520381.2 |
2289 |
Intron |
|
|
XP_011518683.1 |
XM_017018331.1 |
2289 |
Silent Mutation |
GAC,GAT |
D791D |
XP_016873820.1 |
XM_017018332.1 |
2289 |
Silent Mutation |
GAC,GAT |
D753D |
XP_016873821.1 |
XM_017018333.1 |
2289 |
Intron |
|
|
XP_016873822.1 |
View Full Product Details