Product Details

SNP ID
rs201572583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6170163 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATACATGGGCACATGAAGGTTA[C/T]GTTCCATGACAATAACCACTATCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR52B2 PubMed Links

Gene Details

Gene
OR52B2
Gene Name
olfactory receptor family 52 subfamily B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004052.1 164 Missense Mutation NP_001004052.1

View Full Product Details