Product Details

SNP ID
rs201841694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64242424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTTCCGGGTCCTGACAGTACTG[C/T]CCATCTGCCTGAGCGCCGTGGCCAC
Phenotype
MIM: 604189 MIM: 186946 MIM: 600230 MIM: 601140 MIM: 601398
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC4 PubMed Links

Gene Details

Gene
DNAJC4
Gene Name
DnaJ heat shock protein family (Hsp40) member C4
There are no transcripts associated with this gene.

Gene
FKBP2
Gene Name
FK506 binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135208.1 233 Missense Mutation CCC,TCC P13S NP_001128680.1
NM_004470.3 233 Missense Mutation CCC,TCC P13S NP_004461.2
NM_057092.2 233 Missense Mutation CCC,TCC P13S NP_476433.1
XM_005273848.3 233 Missense Mutation CCC,TCC P13S XP_005273905.1
Gene
LOC105369340
Gene Name
uncharacterized LOC105369340
There are no transcripts associated with this gene.

Gene
PLCB3
Gene Name
phospholipase C beta 3
There are no transcripts associated with this gene.

Gene
PPP1R14B
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 14B
There are no transcripts associated with this gene.

Gene
VEGFB
Gene Name
vascular endothelial growth factor B
There are no transcripts associated with this gene.

View Full Product Details