Product Details

SNP ID
rs201124590
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1608162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCCTCCACAGGAGCCACAGCCC[A/C]CCTTGCAGCCCCCACAAGAGCCACA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP5-2 PubMed Links

Gene Details

Gene
KRTAP5-2
Gene Name
keratin associated protein 5-2
There are no transcripts associated with this gene.

Gene
KRTAP5-3
Gene Name
keratin associated protein 5-3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012708.2 Intron NP_001012726.1
Gene
KRTAP5-AS1
Gene Name
KRTAP5-1/KRTAP5-2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details