Product Details
- SNP ID
-
rs200900180
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:57563489 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATTCCCCCCAAACACGACTCAAGA[A/G]CAAGTTTATCATGCATGTGCCATGC
- Phenotype
-
MIM: 602821
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIF5A
PubMed Links
Gene Details
- Gene
- KIF5A
- Gene Name
- kinesin family member 5A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004984.2 |
388 |
Silent Mutation |
GAA,GAG |
E60E |
NP_004975.2 |
View Full Product Details