Product Details

SNP ID
rs200331108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:113427988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCTACCTGGTCTCCAGAATGGA[C/T]GGCCCTGTGGCAGAGCATGCCAAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SDSL PubMed Links

Gene Details

Gene
SDSL
Gene Name
serine dehydratase like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304993.1 650 Silent Mutation GAC,GAT D2D NP_001291922.1
NM_138432.3 650 Silent Mutation GAC,GAT D2D NP_612441.1
XM_005253831.4 650 Silent Mutation GAC,GAT D2D XP_005253888.1
XM_011537846.2 650 Silent Mutation GAC,GAT D2D XP_011536148.1
XM_017018763.1 650 Silent Mutation GAC,GAT D15D XP_016874252.1

View Full Product Details